Assay Details
Gene Symbol: LOC4328210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770954.2 | XP_015626440.1 | 27 - 28 | 3421 | 87 | |
XM_015770955.2 | XP_015626441.1 | 27 - 28 | 3410 | 87 | ||
GenBank mRNA | AK105135.1 | - | 3 - 4 | 418 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
