Assay Details
| Gene Symbol: LOC4328239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768520.2 | XP_015624006.1 | 2 - 3 | 819 | 65 | |
| GenBank mRNA | AK102168.1 | - | 2 - 3 | 705 | 65 | |
More Information
Set Membership: |
|

LOC4328239
Genomic Map