Assay Details
Gene Symbol: LOC4346137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795364.2 | XP_015650850.1 | 1 - 2 | 450 | 96 | |
GenBank mRNA | AB011262.1 | - | 1 - 2 | 357 | 96 | |
AK065141.1 | - | 1 - 2 | 367 | 96 | ||
AK119730.1 | - | 1 - 2 | 368 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
