Assay Details
Gene Symbol: LOC4351811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764858.2 | XP_015620344.1 | 1 - 2 | 240 | 67 | |
GenBank mRNA | AK065655.1 | - | 1 - 2 | 226 | 67 | |
AK102652.1 | - | 1 - 2 | 228 | 67 | ||
AK121853.1 | - | 1 - 2 | 240 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015764858.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
