Assay Details
Gene Symbol: LOC4351811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764858.2 | XP_015620344.1 | 2 - 3 | 2404 | 139 | |
GenBank mRNA | AK065655.1 | - | 2 - 3 | 2390 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
