Assay Details
Gene Symbol: LOC4352214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764655.2 | XP_015620141.1 | 7 - 8 | 781 | 87 | |
GenBank mRNA | AK061316.1 | - | 7 - 8 | 743 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
