Assay Details
Gene Symbol: LOC4352780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765331.2 | XP_015620817.1 | 4 - 5 | 796 | 131 | |
XM_015765332.1 | XP_015620818.1 | 4 - 5 | 795 | 131 | ||
GenBank mRNA | AB118014.1 | - | 3 - 4 | 433 | 131 | |
AK100618.1 | - | 4 - 5 | 796 | 131 | ||
AK105660.1 | - | 4 - 5 | 796 | 131 | ||
FJ940211.1 | - | 4 - 5 | 549 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
