Assay Details
| Gene Symbol: LOC4350053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761367.2 | XP_015616853.1 | 4 - 5 | 908 | 94 | |
| GenBank mRNA | AK061267.1 | - | 5 - 6 | 592 | 94 | |
| AK069330.1 | - | 4 - 5 | 914 | 94 | ||
| AK073964.1 | - | 656 | 94 | |||
| EF122490.1 | - | 4 - 5 | 678 | 94 | ||
| GQ848036.1 | - | 4 - 5 | 631 | 94 | ||
More Information
Set Membership: |
|

LOC4350053
Genomic Map