Assay Details
| Gene Symbol: LOC4350053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761367.2 | XP_015616853.1 | 8 - 9 | 1230 | 61 | |
| GenBank mRNA | AK061267.1 | - | 9 - 10 | 914 | 61 | |
| AK069330.1 | - | 8 - 9 | 1236 | 61 | ||
| AK073964.1 | - | 978 | 61 | |||
| EF122490.1 | - | 8 - 9 | 1000 | 61 | ||
| GQ848036.1 | - | 8 - 9 | 953 | 61 | ||
More Information
Set Membership: |
|

LOC4350053
Genomic Map