Assay Details
Gene Symbol: LOC4350245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762057.2 | XP_015617543.1 | 3 - 4 | 311 | 97 | |
XM_015762058.2 | XP_015617544.1 | 3 - 4 | 311 | 97 | ||
GenBank mRNA | AK102327.1 | - | 3 - 4 | 319 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
