Assay Details
| Gene Symbol: LOC4350646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762534.2 | XP_015618020.1 | 10 - 11 | 2013 | 107 | |
| GenBank mRNA | AK120240.1 | - | 10 - 11 | 2080 | 107 | |
More Information
Set Membership: |
|

LOC4350646
Genomic Map