Assay Details
Gene Symbol: LOC4328422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770433.2 | XP_015625919.1 | 1 - 2 | 308 | 150 | |
XM_015770434.2 | XP_015625920.1 | 1 - 2 | 308 | 150 | ||
XM_015770435.2 | XP_015625921.1 | 1 - 2 | 308 | 150 | ||
GenBank mRNA | AK069161.1 | - | 1 - 2 | 301 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770433.2, XM_015770434.2, XM_015770435.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
