Assay Details
Gene Symbol: LOC4328422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770433.2 | XP_015625919.1 | 8 - 9 | 1341 | 85 | |
XM_015770435.2 | XP_015625921.1 | 8 - 9 | 1341 | 85 | ||
GenBank mRNA | AK069161.1 | - | 7 - 8 | 1508 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
