Assay Details
Gene Symbol: LOC4328422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770433.2 | XP_015625919.1 | 10 - 11 | 1583 | 78 | |
XM_015770434.2 | XP_015625920.1 | 10 - 11 | 1508 | 78 | ||
XM_015770435.2 | XP_015625921.1 | 10 - 11 | 1583 | 78 | ||
GenBank mRNA | AK069161.1 | - | 9 - 10 | 1750 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
