Assay Details
Gene Symbol: LOC4328619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768145.2 | XP_015623631.1 | 5 - 6 | 773 | 101 | |
GenBank mRNA | AK063866.1 | - | 1 - 2 | 80 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
