Assay Details
Gene Symbol: LOC4328619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768145.2 | XP_015623631.1 | 7 - 8 | 885 | 63 | |
GenBank mRNA | AK063866.1 | - | 3 - 4 | 192 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
