Assay Details
Gene Symbol: LOC4328643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769181.2 | XP_015624667.1 | 7 - 8 | 728 | 97 | |
GenBank mRNA | AK068100.1 | - | 7 - 8 | 649 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
