Assay Details
| Gene Symbol: LOC4329063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768898.2 | 3 - 4 | 868 | 117 | ||
| GenBank mRNA | AK106688.1 | - | 4 - 5 | 1137 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4329063| Gene Symbol: LOC4329063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768898.2 | 3 - 4 | 868 | 117 | ||
| GenBank mRNA | AK106688.1 | - | 4 - 5 | 1137 | 117 | |
Set Membership: |
|