Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 12 - 13 | 2038 | 92 | |
GenBank mRNA | AK106285.1 | - | 8 - 9 | 1456 | 92 | |
AY277897.2 | - | 12 - 13 | 1775 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
