Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 16 - 17 | 2623 | 78 | |
GenBank mRNA | AK106581.1 | - | 4 - 5 | 599 | 78 | |
AY277897.2 | - | 16 - 17 | 2360 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
