Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 17 - 18 | 2828 | 126 | |
GenBank mRNA | AK106285.1 | - | 13 - 14 | 2220 | 126 | |
AK106581.1 | - | 5 - 6 | 804 | 126 | ||
AY277897.2 | - | 17 - 18 | 2565 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
