Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 18 - 19 | 3030 | 121 | |
GenBank mRNA | AK106285.1 | - | 14 - 15 | 2422 | 121 | |
AK106581.1 | - | 6 - 7 | 1006 | 121 | ||
AY277897.2 | - | 18 - 19 | 2767 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
