Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 22 - 23 | 3700 | 108 | |
GenBank mRNA | AK106581.1 | - | 10 - 11 | 1676 | 108 | |
AY277897.2 | - | 22 - 23 | 3437 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
