Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 24 - 25 | 3924 | 83 | |
GenBank mRNA | AK106581.1 | - | 12 - 13 | 1900 | 83 | |
AY277897.2 | - | 24 - 25 | 3661 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
