Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 3 - 4 | 653 | 147 | |
GenBank mRNA | AY277897.2 | - | 3 - 4 | 390 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() |
