Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 4 - 5 | 823 | 105 | |
GenBank mRNA | AK106285.1 | - | 1 - 2 | 120 | 105 | |
AY277897.2 | - | 4 - 5 | 560 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
