Assay Details
Gene Symbol: LOC4329409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771294.2 | XP_015626780.1 | 5 - 6 | 945 | 73 | |
GenBank mRNA | AK106285.1 | - | 2 - 3 | 242 | 73 | |
AY277897.2 | - | 5 - 6 | 682 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
