Assay Details
| Gene Symbol: LOC4329892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769896.1 | XP_015625382.1 | 10 - 11 | 1131 | 81 | |
| GenBank mRNA | AF546879.1 | - | 6 - 7 | 573 | 81 | |
| AK062215.1 | - | 10 - 11 | 1115 | 81 | ||
| AK099086.1 | - | 10 - 11 | 1100 | 81 | ||
| BT131063.1 | - | 5 - 6 | 539 | 81 | ||
| GQ848032.1 | - | 10 - 11 | 921 | 81 | ||
| KY038497.1 | - | 10 - 11 | 956 | 81 | ||
More Information
Set Membership: |
|

LOC4329892
Genomic Map