Assay Details
| Gene Symbol: LOC4329905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767821.2 | XP_015623307.1 | 4 - 5 | 454 | 87 | |
| GenBank mRNA | AK068016.1 | - | 4 - 5 | 417 | 87 | |
| AK068109.1 | - | 4 - 5 | 435 | 87 | ||
More Information
Set Membership: |
|

LOC4329905
Genomic Map