Assay Details
Gene Symbol: LOC4330297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770130.2 | XP_015625616.1 | 6 - 7 | 1963 | 63 | |
GenBank mRNA | AB101648.1 | - | 6 - 7 | 1649 | 63 | |
AK111544.1 | - | 6 - 7 | 1969 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
