Assay Details
Gene Symbol: LOC4330484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769939.2 | XP_015625425.1 | 22 - 23 | 4514 | 95 | |
XM_015769940.2 | XP_015625426.1 | 21 - 22 | 4277 | 95 | ||
GenBank mRNA | AB477099.1 | - | 21 - 22 | 4200 | 95 | |
AK064381.1 | - | 3 - 4 | 625 | 95 | ||
AK121261.1 | - | 22 - 23 | 4514 | 95 | ||
AK121875.1 | - | 22 - 23 | 4469 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
