Assay Details
| Gene Symbol: LOC4330726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771971.2 | XP_015627457.1 | 1 - 2 | 415 | 70 | |
| XM_015771973.2 | XP_015627459.1 | 1 - 2 | 380 | 70 | ||
| XM_015771978.2 | XP_015627464.1 | 1 - 2 | 415 | 70 | ||
| XM_015771979.2 | XP_015627465.1 | 1 - 2 | 415 | 70 | ||
| XM_015771980.2 | XP_015627466.1 | 1 - 2 | 415 | 70 | ||
| GenBank mRNA | AK101365.1 | - | 1 - 2 | 380 | 70 | |
| AK111248.1 | - | 1 - 2 | 366 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771973.2 transcripts.
Set Membership: |
|

LOC4330726
Genomic Map