Assay Details
Gene Symbol: LOC4330726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771971.2 | XP_015627457.1 | 2 - 3 | 519 | 98 | |
XM_015771972.2 | XP_015627458.1 | 2 - 3 | 510 | 98 | ||
GenBank mRNA | AK101365.1 | - | 2 - 3 | 484 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
