Assay Details
Gene Symbol: LOC4331050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769665.2 | XP_015625151.1 | 5 - 6 | 818 | 150 | |
GenBank mRNA | AB037155.1 | - | 5 - 6 | 769 | 150 | |
AK062185.1 | - | 5 - 6 | 785 | 150 | ||
AK067051.1 | - | 5 - 6 | 685 | 150 | ||
AK101474.1 | - | 5 - 6 | 812 | 150 | ||
AK103936.1 | - | 5 - 6 | 798 | 150 | ||
D17788.1 | - | 5 - 6 | 768 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
