Assay Details
Gene Symbol: LOC4331151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769947.2 | XP_015625433.1 | 2 - 3 | 422 | 111 | |
GenBank mRNA | AK068473.1 | - | 2 - 3 | 369 | 111 | |
AK100685.1 | - | 2 - 3 | 368 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769947.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
