Assay Details
| Gene Symbol: LOC4331151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769947.2 | XP_015625433.1 | 4 - 5 | 1426 | 70 | |
| GenBank mRNA | AK068473.1 | - | 4 - 5 | 1373 | 70 | |
| AK100685.1 | - | 4 - 5 | 1372 | 70 | ||
More Information
Set Membership: |
|

LOC4331151
Genomic Map