Assay Details
Gene Symbol: LOC4331151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769947.2 | XP_015625433.1 | 6 - 7 | 200 | ||
GenBank mRNA | AK100685.1 | - | 5 - 6 | 1490 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
