Assay Details
Gene Symbol: LOC4331151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769947.2 | XP_015625433.1 | 7 - 8 | 1862 | 66 | |
GenBank mRNA | AK100685.1 | - | 6 - 7 | 1728 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
