Assay Details
Gene Symbol: LOC4331151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769947.2 | XP_015625433.1 | 9 - 10 | 2215 | 81 | |
GenBank mRNA | AK068473.1 | - | 7 - 8 | 2759 | 81 | |
AK100685.1 | - | 8 - 9 | 2081 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
