Assay Details
| Gene Symbol: LOC4331279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775282.2 | XP_015630768.1 | 6 - 7 | 2350 | 137 | |
| GenBank mRNA | AK068556.1 | - | 6 - 7 | 2348 | 137 | |
| AK121880.1 | - | 6 - 7 | 2341 | 137 | ||
More Information
Set Membership: |
|

LOC4331279
Genomic Map