Assay Details
| Gene Symbol: LOC4331292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776507.2 | XP_015631993.1 | 7 - 8 | 849 | 121 | |
| XM_015776508.2 | XP_015631994.1 | 7 - 8 | 842 | 121 | ||
| GenBank mRNA | AK072180.1 | - | 7 - 8 | 827 | 121 | |
| AK098927.1 | - | 7 - 8 | 763 | 121 | ||
More Information
Set Membership: |
|

LOC4331292
Genomic Map