Assay Details
| Gene Symbol: LOC4331993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772542.2 | XP_015628028.1 | 2 - 3 | 1256 | 86 | |
| XM_015772544.2 | XP_015628030.1 | 2 - 3 | 1036 | 86 | ||
| GenBank mRNA | AK103674.1 | - | 1 - 2 | 534 | 86 | |
| AK105404.1 | - | 1 - 2 | 182 | 86 | ||
More Information
Set Membership: |
|

LOC4331993
Genomic Map