Assay Details
| Gene Symbol: LOC4331993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772542.2 | XP_015628028.1 | 3 - 4 | 1325 | 63 | |
| XM_015772544.2 | XP_015628030.1 | 3 - 4 | 1105 | 63 | ||
| GenBank mRNA | AK103674.1 | - | 2 - 3 | 603 | 63 | |
| AK105404.1 | - | 2 - 3 | 251 | 63 | ||
More Information
Set Membership: |
|

LOC4331993
Genomic Map