Assay Details
| Gene Symbol: LOC4332128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774530.2 | XP_015630016.1 | 14 - 15 | 2674 | 120 | |
| GenBank mRNA | AK242575.1 | - | 14 - 15 | 2680 | 120 | |
More Information
Set Membership: |
|

LOC4332128
Genomic Map