Assay Details
| Gene Symbol: LOC4332994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772525.2 | XP_015628011.1 | 1 - 2 | 381 | 82 | |
| GenBank mRNA | AK066831.1 | - | 1 - 2 | 337 | 82 | |
| AK072987.1 | - | 1 - 2 | 363 | 82 | ||
| AK073780.1 | - | 1 - 2 | 337 | 82 | ||
More Information
Set Membership: |
|

LOC4332994
Genomic Map