Assay Details
Gene Symbol: LOC4333431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776969.2 | XP_015632455.1 | 1 - 2 | 123 | 122 | |
GenBank mRNA | AK065981.1 | - | 1 - 2 | 87 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776969.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
