Assay Details
| Gene Symbol: LOC4333983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777179.2 | XP_015632665.1 | 12 - 13 | 1722 | 79 | |
| GenBank mRNA | AK068068.1 | - | 12 - 13 | 1720 | 79 | |
| AK121524.1 | - | 3 - 4 | 285 | 79 | ||
More Information
Set Membership: |
|

LOC4333983
Genomic Map