Assay Details
Gene Symbol: LOC4333983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777179.2 | XP_015632665.1 | 13 - 14 | 1750 | 125 | |
GenBank mRNA | AK068068.1 | - | 13 - 14 | 1748 | 125 | |
AK121524.1 | - | 4 - 5 | 313 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
