Assay Details
| Gene Symbol: LOC4333983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777179.2 | XP_015632665.1 | 1 - 2 | 419 | 64 | |
| GenBank mRNA | AK068068.1 | - | 1 - 2 | 417 | 64 | |
More Information
Set Membership: |
|

LOC4333983
Genomic Map