Assay Details
Gene Symbol: LOC4333983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777179.2 | XP_015632665.1 | 3 - 4 | 680 | 88 | |
GenBank mRNA | AK068068.1 | - | 3 - 4 | 678 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
